Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VGuard™ Bouffant Caps

Catalog No. 17005953
Encompass
Encompass_Preferred
Change view
Click to view available options
Color:
Green
Red
White
Style:
Pancake
Pleated
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Style
17-005-953 White Pleated
17-005-943 White Pancake
17-005-944 White Pancake
17-005-945 White Pancake
17-005-948 - Pancake
17-005-949 Red Pancake
17-005-951 Green Pancake
17-005-952 White Pleated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 17-005-953 Supplier VGuard™ Supplier No. B11B12
May include imposed supplier surcharges.
Add to Cart
Add to Cart
This item is not returnable. View return policy

Breathable, soft, and lightweight protection when full hair coverage is required.

  • Strong, elastic headband
  • Superior barrier properties
  • Available in pancake or pleated styles

Specifications

Material Spunbond Polypropylene
Color White
Disposable Single-use
Model B11B12
For Use With (Application) Food Service, Food Processing, Industrial, Janitorial, Manufacturing
Style Pleated
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.