Learn More
Polysciences, Inc. Picric acid, formalin and acetic acid fixative. Bouin's fixative is excellent for use in preserving soft and delicate tissue structures.
SDP
Catalog No. NC0726856
Encompass
Picric acid, formalin and acetic acid fixative. Bouin's fixative is excellent for use in preserving soft and delicate tissue structures.
Catalog No. NC0726856 Supplier Polysciences, Inc. Supplier No. 160451
Picric acid, formalin and acetic acid fixative. Bouin's fixative is excellent for use in preserving soft and delicate tissue structures.