Learn More
Bouin′s Solution, 250 mL, G-Biosciences
SCI_ED
Catalog No. S001143
Change view
Click to view available options
Quantity:
250 mL
Catalog No. Quantity
S001143 250 mL
1 options
Catalog No. S001143 Supplier G-Biosciences Supplier No. 7862244
Add to Cart
Add to Cart
Bouin’s solution is used as a mordant in various trichrome staining.
Uses:
  • Fixing soft and delicate tissues.
  • Fixing gastrointestinal tract biopsies
  • Fixing tissues with chromosomes in mitosis and meiosis.
Specifications
Quantity 250 mL
Product Type Bouin′s Solution