Learn More
IMEB, Inc Bouin's Fixative Substitute, No Picric Acid, Pre-filled 3 Oz, 96 per Case
SDP
Catalog No. NC0457974
Encompass
A low-hazard replacement for Bouin’s Fixative, this substitute provides all the benefits of Bouin’s without the use of picric acid. Available in a variety of sizes and pre-filled specimen containers. Pre-filled 3 Oz, 96 per Case
Catalog No. NC0457974 Supplier IMEB, Inc Supplier No. BFSP3
A low-hazard replacement for Bouin’s Fixative, this substitute provides all the benefits of Bouin’s without the use of picric acid. Available in a variety of sizes and pre-filled specimen containers. Pre-filled 3 Oz, 96 per Case