Learn More
IMEB, Inc Bouin's Fixative 1 Gallon
SDP
Catalog No. NC9178427
Encompass_Preferred
Bouin’s fixative is especially good for gastrointestinal tract biopsies because it allows crisper and better nuclear staining than 10% neutral-buffered formalin. It is a good fixative when tissue structure with a soft and delicate texture must be preserved. 1 Gallon Bottle. Available in a variety of other sizes and pre-filled specimen containers. Substitute solution also available.
Catalog No. NC9178427 Supplier IMEB, Inc Supplier No. S129
Bouin’s fixative is especially good for gastrointestinal tract biopsies because it allows crisper and better nuclear staining than 10% neutral-buffered formalin. It is a good fixative when tissue structure with a soft and delicate texture must be preserved. 1 Gallon Bottle. Available in a variety of other sizes and pre-filled specimen containers. Substitute solution also available.