Learn More
IMEB, Inc Bouin's Fixative Substitute with No Picrid Acid, 32 oz Bottle
SDP
Catalog No. NC9772216
Encompass_Preferred
A low-hazard replacement for Bouin’s Fixative, this substitute provides all the benefits of Bouin’s without the use of picric acid. 32 oz bottle. Also available for purchase in a variety of sizes and pre-filled specimen containers.
Catalog No. NC9772216 Supplier IMEB, Inc Supplier No. BFS32
A low-hazard replacement for Bouin’s Fixative, this substitute provides all the benefits of Bouin’s without the use of picric acid. 32 oz bottle. Also available for purchase in a variety of sizes and pre-filled specimen containers.