Learn More
Electron Microscopy Sciences Bouin's Fluid, 500ml
SDP
Catalog No. 5030098
Encompass_Preferred
Goldner's Trichrome Method For connective tissue stains-Elastic tissue stain. Stain with a poly-acid step. Stain results:Nuclear chromatin: Brownish-Black, Cytoplasm: Bright Red, Erythrocytes: Orange, Muscle: Red, Collagen: Green
Catalog No. 50-300-98 Supplier Electron Microscopy Sciences Supplier No. 2638601
Goldner's Trichrome Method For connective tissue stains-Elastic tissue stain. Stain with a poly-acid step. Stain results:Nuclear chromatin: Brownish-Black, Cytoplasm: Bright Red, Erythrocytes: Orange, Muscle: Red, Collagen: Green