Learn More
Newcomer Supply Bouin Fluid, 1 gallon
SDP
Catalog No. NC0827069
Encompass
Newcomer Supply Bouin Fluid is a ready-to-use picric acid based fixative combined with acetic acid and formalin.  Bouin Fluid penetrates rapidly, fixes evenly, provides crisp nuclear staining and preserves structures with soft and delicate features. Shelf Life is 2 years from date of manufacture. Store at 15-30°C. Bouin Fluid is recommended for a variety of specimens including bone marrow clots and biopsies, gastrointestinal tract biopsies, testicular biopsies and lymph nodes. It also serves as both fixative and mordant for tissues stained with trichrome procedures.
Catalog No. NC0827069 Supplier Newcomer Supply Supplier No. 1020B
Newcomer Supply Bouin Fluid is a ready-to-use picric acid based fixative combined with acetic acid and formalin.  Bouin Fluid penetrates rapidly, fixes evenly, provides crisp nuclear staining and preserves structures with soft and delicate features. Shelf Life is 2 years from date of manufacture. Store at 15-30°C. Bouin Fluid is recommended for a variety of specimens including bone marrow clots and biopsies, gastrointestinal tract biopsies, testicular biopsies and lymph nodes. It also serves as both fixative and mordant for tissues stained with trichrome procedures.