Learn More
Enzo Life Sciences Bourgeonal (25mg). CAS: 18127-01-0
SDP
Supplier:  Enzo Life Sciences BMLN1560025
Encompass_Preferred
A previously undescribed human testicular olfactory receptor known as hOR17-4 was cloned and functionally expressed. Bourgeonal is a potent agonist at hOR17-4 and acts as a strong chemoattractant in sperm behavioral assays. The bourgeonal, hOR17-4 signaling pathway potentially governs chemical communication between sperm and egg and may be used to manipulate or block fertilization. Alternative name: 4-t-Butylbenzenepropionaldehyde. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (25 mg/ml)or ethanol (25 mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-1173