Learn More
MSC Bouton Lens Cleaning Station, Anti-Fog Anti-Static, Silicone-Free, 8 Stations/Case
Catalog No. 5000793137
Encompass
This lens cleaning station is designed for easy access and mobility, suitable for various work environments. It features a disposable dispenser and is effective for cleaning plastic, glass, and polycarbonate lenses. The station can also be conveniently mounted on walls.
  • Disposable dispenser
  • Contains 8 oz. of anti-fog/anti-static cleaner
  • Includes two boxes of 300-count tissues
  • Spray pump bottle
  • Lint-free absorbent tissues made from 100% recycled materials
  • Suitable for plastic, glass, or polycarbonate lenses
  • Wall-mountable
  • Silicone-free
Catalog No. 50-007-93137 Supplier MSC Supplier No. 58613357
This lens cleaning station is designed for easy access and mobility, suitable for various work environments. It features a disposable dispenser and is effective for cleaning plastic, glass, and polycarbonate lenses. The station can also be conveniently mounted on walls.
  • Disposable dispenser
  • Contains 8 oz. of anti-fog/anti-static cleaner
  • Includes two boxes of 300-count tissues
  • Spray pump bottle
  • Lint-free absorbent tissues made from 100% recycled materials
  • Suitable for plastic, glass, or polycarbonate lenses
  • Wall-mountable
  • Silicone-free