Learn More
MSC Bouton Safety Glasses, Anti-Fog, Scratch-Resistant, Clear Mirror, Frameless, 1 ea
Catalog No. 5000794393
Encompass
These safety glasses feature anti-fog and scratch-resistant clear mirror polycarbonate lenses, designed for indoor/outdoor use. With a frameless, wrap-around design and non-adjustable temples, they provide eye protection and meet ANSI Z87.1+ standards.
  • Anti-fog and scratch-resistant lenses
  • Clear mirror polycarbonate lenses
  • Frameless, wrap-around design
  • Non-adjustable temples
  • Universal size
  • UV protection
  • Meets ANSI Z87.1+ standards
  • Foam padding for comfort
Catalog No. 50-007-94393 Supplier MSC Supplier No. 48073381
These safety glasses feature anti-fog and scratch-resistant clear mirror polycarbonate lenses, designed for indoor/outdoor use. With a frameless, wrap-around design and non-adjustable temples, they provide eye protection and meet ANSI Z87.1+ standards.
  • Anti-fog and scratch-resistant lenses
  • Clear mirror polycarbonate lenses
  • Frameless, wrap-around design
  • Non-adjustable temples
  • Universal size
  • UV protection
  • Meets ANSI Z87.1+ standards
  • Foam padding for comfort