Learn More
MSC Bouton Safety Glasses, Zenon Ultra-Lyte, Clear Lens, Anti-Fog, 1 ea
Catalog No. 5000792298
Encompass
These rimless safety glasses offer an ultra-lightweight design for all-day wear, conforming to the face to minimize gaps and ensure a comfortable fit. They feature a squared lens shape with a modern taper and an advanced anti-fog coating for clear vision in demanding environments.
  • Squared lens shape with a slight taper for modern styling
  • Ultra-lightweight design that conforms to the head for all-day wear
  • Spring fit temples for a comfortable and conforming fit
  • FogLess® 3Sixty™ enhanced hydrophobic action resists lens fogging and ensures clear vision
  • Molded nose bridge designed to fit most users
  • Features a 10 base curve lens
  • Meets ANSI Z87.1+ -2020 standards
Catalog No. 50-007-92298 Supplier MSC Supplier No. 77789501
These rimless safety glasses offer an ultra-lightweight design for all-day wear, conforming to the face to minimize gaps and ensure a comfortable fit. They feature a squared lens shape with a modern taper and an advanced anti-fog coating for clear vision in demanding environments.
  • Squared lens shape with a slight taper for modern styling
  • Ultra-lightweight design that conforms to the head for all-day wear
  • Spring fit temples for a comfortable and conforming fit
  • FogLess® 3Sixty™ enhanced hydrophobic action resists lens fogging and ensures clear vision
  • Molded nose bridge designed to fit most users
  • Features a 10 base curve lens
  • Meets ANSI Z87.1+ -2020 standards