Learn More
MSC Bouton Xtricate Safety Glasses, Clear FogLess 3Sixty Anti-Fog, 1 Pair
Catalog No. 5000769592
Encompass
These Xtricate safety glasses feature a comfortable design and sporty styling with advanced fog protection.
  • Rubber nose pads offer a more secure fit
  • Lightweight, semi-rimless design provides all-day user comfort
  • Flexible temples provide a secure fit without uncomfortable pressure points
  • FogLess® 3Sixty™ enhanced hydrophobic action resists lens fogging and provides clear vision in extreme working conditions
  • An 8.5 base curve lens
  • Meets ANSI Z87.1-2015 standards
Catalog No. 50-007-69592 Supplier MSC Supplier No. 28498962
These Xtricate safety glasses feature a comfortable design and sporty styling with advanced fog protection.
  • Rubber nose pads offer a more secure fit
  • Lightweight, semi-rimless design provides all-day user comfort
  • Flexible temples provide a secure fit without uncomfortable pressure points
  • FogLess® 3Sixty™ enhanced hydrophobic action resists lens fogging and provides clear vision in extreme working conditions
  • An 8.5 base curve lens
  • Meets ANSI Z87.1-2015 standards