Learn More
MSC Bouton Zenon Z12 Rimless Safety Glasses, Clear Anti-Scratch, 1 ea
Catalog No. 5000677224
Encompass
These Zenon Z12 safety glasses feature a lightweight, rimless design with clear anti-scratch coated lenses, suitable for various industrial applications.
  • Lightweight, rimless design provides all-day user comfort.
  • Thin, flexible temples provide a secure fit without uncomfortable pressure points.
  • Molded nose bridge design to fit the vast majority of users.
  • Features a 9.5 base curve lens.
  • Meets ANSI Z87.1+.
  • Meets CSA Z94.3.
Catalog No. 50-006-77224 Supplier MSC Supplier No. 16297640
These Zenon Z12 safety glasses feature a lightweight, rimless design with clear anti-scratch coated lenses, suitable for various industrial applications.
  • Lightweight, rimless design provides all-day user comfort.
  • Thin, flexible temples provide a secure fit without uncomfortable pressure points.
  • Molded nose bridge design to fit the vast majority of users.
  • Features a 9.5 base curve lens.
  • Meets ANSI Z87.1+.
  • Meets CSA Z94.3.