Learn More
Cayman Chemical BovIn Catlase 1ea
SDP
Catalog No. 502893791
Encompass_Preferred
An internal standard for the quantification of diazinon by GC- or LC-MS
Catalog No. 50-289-3791 Supplier Cayman Chemical Supplier No. 6000541 EA
An internal standard for the quantification of diazinon by GC- or LC-MS