Learn More
Cayman Chemical BovIn Heart MItochondrIa100uL
SDP
Catalog No. 502892868
Encompass_Preferred
A GnRH receptor antagonist (Kis 6 2.2 and 3.8 nM for the human mouse and rat receptors respectively) selective for the GnRH receptor over a panel of 43 receptors ion channels and enzymes at 1 UM prevents GnRH-induced increases in inositol phosphate production in HEK293 cells expressing the human GnRH receptor (KB 25 nM) oral or intravenous administration decreases plasma LH levels in castrated rats inhibits GnRH-A-induced increases in plasma LH and testosterone levels in non-castrated rats at 20 mg/kg
Catalog No. 50-289-2868 Supplier Cayman Chemical Supplier No. 700019100 UL
Add to Cart
Add to Cart
A GnRH receptor antagonist (Kis 6 2.2 and 3.8 nM for the human mouse and rat receptors respectively) selective for the GnRH receptor over a panel of 43 receptors ion channels and enzymes at 1 UM prevents GnRH-induced increases in inositol phosphate production in HEK293 cells expressing the human GnRH receptor (KB 25 nM) oral or intravenous administration decreases plasma LH levels in castrated rats inhibits GnRH-A-induced increases in plasma LH and testosterone levels in non-castrated rats at 20 mg/kg