Learn More
Ubpbio BOVINE 19S (PA700), 25UG
Supplier:  Ubpbio A1300
Encompass_Preferred
Proteasomes/Activators/Subunits – Bovine 19S complex (UBPBio catalog# A1300, A1301) - Purified from bovine red blood cells. The 19S regulatory particle (PA700) locates on either one or both ends of the 20S core particle of the 26S proteasome. It has 19 subunits including six AAA ATPases that associate with α subunits of the 20S proteasome to form the 26S proteasome. PA700 plays an essential role in processing substrate proteins to be degraded by the 20S proteasome, including binding, deubiquitinating and unfolding polyubiquitinated proteins.
Catalog No. 50-112-6848