Learn More
American Research Products Inc BoVine a1 Acid glycoprotein a
Catalog No. 5000329683
Encompass
BoVine a1-Acid glycoprotein a1-AGP ELISA Kit from CUSABIO Detection Range 62 5 ng/mL-4000 ng/mL Sensitivity 15 6 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-29683 Supplier American Research Products Inc Supplier No. CSBE08579B
BoVine a1-Acid glycoprotein a1-AGP ELISA Kit from CUSABIO Detection Range 62 5 ng/mL-4000 ng/mL Sensitivity 15 6 ng/mL Sample serum plasma tissue homogenates Method Competitive