Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bovine Activated Protein C (Dansyl-EGR-CK) Native Protein

Catalog No. PIRP43098 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP43098 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP43098 Supplier Invitrogen™ Supplier No. RP43098
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Native Protein

RP-43098 contains native bovine Activated Protein C (Dansyl-EGR-CK) protein. RP-43098 may be used as a positive control in many immunological assays.

Protein C is a vitamin K-dependent plasma glycoprotein that is cleaved to its activated form by the thrombin-thrombomodulin complex. Protein C in its activated form contains a serine protease domain and functions in degradation of the activated forms of coagulation factors V and VIII. Protein C is a vitamin K-dependent serine protease produced in the liver and made up of 2 polypeptide chains. The 62 kDa proenzyme is activated by thrombin bound to an endothelial surface receptor and the active enzyme cleaves factor Va and VIIIa and thus inhibits blood coagulation. The molecular weight of the active enzyme is 55 kDa and the normal concentrations in human plasma is approximately 1-3 ng/mL because of the very fast turnover, the proenzyme concentration is approximately 3 μg/mL. Mutations in the Protein C gene have been associated with thrombophilia due to protein C deficiency, neonatal purpura fulminans, and recurrent venous thrombosis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P00745
Concentration 3.2 mg/mL
For Use With (Application) Neutralization
Formulation 50% water with no preservative
Gene ID (Entrez) 281428
Molecular Weight (g/mol) 52.65 kDa
Name Bovine Activated Protein C (Dansyl-EGR-CK)
pH Range 7.4
Purification Method SDS-PAGE
Quantity 50 μg
Source Bovine
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias Activation peptide; Anticoagulant protein C; APC; autoprothrombin IIA; Blood coagulation factor XIV; EDQVDPRLIDGK; inactivator of coagulation factors Va, VIII; PC; prepro-protein C; Proc; PROC1; protein C; protein C (inactivator of coagulation factors Va and VIIIa); protein C prepropeptide; protein C, inactivator of coagulation factors Va and VIIIa; THPH3; THPH4; type I protein C; vitamin K-dependent protein C; Vitamin K-dependent protein C heavy chain; Vitamin K-dependent protein C light chain
Common Name Protein C
Gene Symbol PROC
Product Type Protein
Conjugate Unconjugated
Species Bovine
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Form Liquid
Protein Subtype Native
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.