Learn More
Biomatik Corporation Bovine Adiponectin, Adp Elisa Kit
Supplier:  Biomatik Corporation EKC31538
Encompass_Preferred
Bovine Adiponectin, Adp Elisa Kit for detecting Adiponectin, C1Q And Collagen Domain Containing in serum, plasma, tissue homogenates. Species Reactivity: Bovine (Bos taurus). Detection Range: 3.125-50ug/ml. Sensitivity: 3.125ug/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: Q3Y5Z3; ADIPOQ. Target Synonyms: ADIPOQ; ACRP30; APM1Adiponectin; 30 kDa adipocyte complement-related protein; Adipocyte complement-related 30 kDa protein; ACRP30; Adipocyte; C1q and collagen domain-containing protein; Adipose most abundant gene transcript 1 protein; apM-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4613