Learn More
American Research Products Inc BoVine adrenocorticotropic hor
Catalog No. 5000330069
Encompass
BoVine adrenocorticotropic hormone (ACTH) ELISA kit from CUSABIO Detection Range 7 8 pg/ml-500 pg/ml Sensitivity 3 9 pg/ml Sample serum plasma Method Competitive
Catalog No. 50-003-30069 Supplier American Research Products Inc Supplier No. CSBE14322B
BoVine adrenocorticotropic hormone (ACTH) ELISA kit from CUSABIO Detection Range 7 8 pg/ml-500 pg/ml Sensitivity 3 9 pg/ml Sample serum plasma Method Competitive