Learn More
American Research Products Inc BoVine Albumin Alb ELISA Kit
Catalog No. 5000326155
Encompass
BoVine Albumin (Alb) ELISA Kit from CUSABIO Detection Range 0 78 g/mL-50 g/mL Sensitivity 0 39 g/mL Sample serum plasma Method Competitive
Catalog No. 50-003-26155 Supplier American Research Products Inc Supplier No. CSBE08664BIS
BoVine Albumin (Alb) ELISA Kit from CUSABIO Detection Range 0 78 g/mL-50 g/mL Sensitivity 0 39 g/mL Sample serum plasma Method Competitive