Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Bovine Albumin Cohn

Catalog No. MP8810013 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP8810013 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP8810013 Supplier MP Biomedicals™ Supplier No. 08810013
Add to Cart
Add to Cart

Heat shocked, precipitated by cold ethanol and crystallized.

  • Used to solubilize lipids, and used as blocking agent in Western blots or ELISA applications
TRUSTED_SUSTAINABILITY

Specifications

Concentration 98% protein
For Use With (Application) Immunoassay, Mammalian Cell Culture
Name Bovine Albumin Cohn
Preparation Method Heat shocked, precipitated by cold ethanol and crystallized
Quantity 10 g
Storage Requirements Store at 2° to 8°C
Form Crystalline
Protein Subtype Albumin
Purity or Quality Grade >99%
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.