Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Bovine Albumin Fraction V (7.5% solution)
GREENER_CHOICE

Catalog No. 501215315
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-121-5315 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-121-5315 Supplier Gibco™ Supplier No. 15260037
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Bovine Albumin Fraction V (7.5% solution)

7.5 g/100 mL

Prepared with Bovine Serum Albumin Fraction V in phosphate-buffered saline.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in refrigerator (2–8°C).
Cell Type Mammalian Cells
Form Liquid
Product Type BSA
pH 6 to 8
Quantity 100 mL
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.