Learn More
Biomatik Corporation Bovine Albumin (Alb) Elisa Kit
Supplier:  Biomatik Corporation EKC31001
Encompass_Preferred
Bovine Albumin (Alb) Elisa Kit for detecting Albumin in serum, plasma, tissue homogenates. Species Reactivity: Bovine (Bos taurus). Detection Range: 0.313-80ug/ml. Sensitivity: 0.148ug/mL. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P02769; ALB. Target Synonyms: ALB; Serum albumin; BSA; allergen Bos d 6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9202