Learn More
Biomatik Corporation Bovine Alkaline Phosphatase (ALP) CLIA Kit
Supplier:  Biomatik Corporation EKU08942
Encompass_Preferred
Bovine Alkaline Phosphatase (ALP) CLIA Kit for detecting ALP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 68.6-50000pg/ml. Sensitivity: 25.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P09487. Gene ID: 280994. Target Synonyms: AKP; ALKP; Basic Phosphatase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6123