Learn More
American Research Products Inc BoVine Alpha 2 HS glycoprotein
Catalog No. 5000327552
Encompass
BoVine Alpha-2-HS-glycoprotein (AHSG) ELISA kit from CUSABIO Detection Range 125 ng/mL-8000 ng/mL Sensitivity 31 25 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-27552 Supplier American Research Products Inc Supplier No. CSBEL001485BO
BoVine Alpha-2-HS-glycoprotein (AHSG) ELISA kit from CUSABIO Detection Range 125 ng/mL-8000 ng/mL Sensitivity 31 25 ng/mL Sample serum plasma tissue homogenates Method Competitive