Learn More
American Research Products Inc BoVine Alpha 2 macroglobulin
Catalog No. 5000328373
Encompass
BoVine Alpha-2-macroglobulin (A2M) ELISA kit from CUSABIO Detection Range 0 312 g/mL- 20 g/mL Sensitivity 0 078 g/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-28373 Supplier American Research Products Inc Supplier No. CSBEL001004BO
BoVine Alpha-2-macroglobulin (A2M) ELISA kit from CUSABIO Detection Range 0 312 g/mL- 20 g/mL Sensitivity 0 078 g/mL Sample serum plasma tissue homogenates Method Competitive