Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Bovine ALPI native Protein from calf intestine

Catalog No. 89933231 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
5000 Units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-231 5000 Units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-231 Supplier Abnova™ Supplier No. P5337
Add to Cart
Add to Cart

Used for Enzyme, SDS-PAGE

Specifications

For Use With (Application) Enzyme Activity, SDS-PAGE
Formulation Lyophilized from Tris buffer containing Mg and Zn
Gene ID (Entrez) 280993
Name ALPI (Bovine) Native Protein
Preparation Method Native protein purified from calf intestine
Quantity 5000 Units
Storage Requirements Store at -20°C. For best storage after opening, redesiccate over silica gel under vacuum before returning to -20°C.
Regulatory Status RUO
Gene Alias IAP
Common Name ALPI
Gene Symbol ALPI
Biological Activity 1,000 units/mg of protein. Unit definition: That amount of enzyme catalyzing the liberation of 1μmole p-nitrophenol per minute at 37°C and pH 9.8 in DEA buffer.
Species Bovine
Protein Tag None
Expression System Native protein purified from calf intestine
Form Lyophilized
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.