Learn More
American Research Products Inc BoVine Angiopoietin 1 ANGPT1
Catalog No. 5000328377
Encompass
BoVine Angiopoietin-1 (ANGPT1) ELISA kit from CUSABIO Detection Range 12 5 ng/mL-800 ng/mL Sensitivity 1 384 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28377 Supplier American Research Products Inc Supplier No. CSBEL001706BO
BoVine Angiopoietin-1 (ANGPT1) ELISA kit from CUSABIO Detection Range 12 5 ng/mL-800 ng/mL Sensitivity 1 384 ng/mL Sample serum plasma tissue homogenates Method Sandwich