Learn More
Biomatik Corporation Bovine Angiotensin II (AngII) ELISA Kit
Supplier:  Biomatik Corporation EKU02402
Encompass_Preferred
Bovine Angiotensin II (AngII) ELISA Kit for detecting AngII in serum, plasma, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 24.69-2000pg/ml. Sensitivity: 8.46pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01017. Target Synonyms: Ang-II; Angiotensin-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5878