Learn More
ILEX LIFE SCIENCES LLC Apo Transferrin Bovine, Native Protein (Serum), 50 mg
SDP
Catalog No. 502339485
Encompass_Preferred
Apo Transferrin Bovine, Native Protein (Serum) is a glycoprotein purified from bovine serum that has a molecular mass of approximately 77 kDa. It has a low iron content of < 40 ppm. FORMULATION: Sterile filtered off-white lyophilized (freeze-dried) powder. The protein (1 mg/ml) was lyophilized with no additives. PURITY: Greater than 95.0% as determined by SDS-PAGE. IRON CONTENT: The Iron content was estimated by ICP-OES and was found to be less than 40 ppm. OTHER NAMES: Bovine ApoTransferrin, iron-free transferrin, Serotransferrin, Transferrin, Siderophilin, Beta-1-metal-binding globulin, TF, PRO1557, PRO2086, DKFZp781D0156, ATF.
Catalog No. 50-233-9485 Supplier ILEX LIFE SCIENCES LLC Supplier No. A15110050
Apo Transferrin Bovine, Native Protein (Serum) is a glycoprotein purified from bovine serum that has a molecular mass of approximately 77 kDa. It has a low iron content of < 40 ppm. FORMULATION: Sterile filtered off-white lyophilized (freeze-dried) powder. The protein (1 mg/ml) was lyophilized with no additives. PURITY: Greater than 95.0% as determined by SDS-PAGE. IRON CONTENT: The Iron content was estimated by ICP-OES and was found to be less than 40 ppm. OTHER NAMES: Bovine ApoTransferrin, iron-free transferrin, Serotransferrin, Transferrin, Siderophilin, Beta-1-metal-binding globulin, TF, PRO1557, PRO2086, DKFZp781D0156, ATF.