Learn More
Biomatik Corporation Bovine Apolipoprotein A4 (APOA4) ELISA Kit
Supplier:  Biomatik Corporation EKU08308
Encompass_Preferred
Bovine Apolipoprotein A4 (APOA4) ELISA Kit for detecting APOA4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.69ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q32PJ2. Gene ID: 537301. Target Synonyms: ApoA-IV; ApoAIV; Apo-A4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7327