Learn More
Biomatik Corporation Bovine Aprotinin (AP) CLIA Kit
Supplier:  Biomatik Corporation EKU08651
Encompass_Preferred
Bovine Aprotinin (AP) CLIA Kit for detecting AP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 1.95-500ng/ml. Sensitivity: 0.71ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Accession Number: P00974. Target Synonyms: BPTI; Trasylol; Pancreatic Trypsin Inhibitor; Basic protease inhibitor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6477