Learn More
Bioworld BOVINE APROTININ
Supplier:  Bioworld 220600132
Encompass_Preferred
Recombinant Bovine Aprotinin (AP) Source: Maize (Corn) Background: Aprotinin inhibits the activity of several proteolytic enzymes such as chymotrypsin, kallikrein, plasmin and trypsin. It is present in blood and in most tissues, wi...
Catalog No. 50-253-107