Learn More
Biomatik Corporation Bovine Aspartate Aminotransferase (AST) ELISA Kit
Supplier:  Biomatik Corporation EKU02591
Encompass_Preferred
Bovine Aspartate Aminotransferase (AST) ELISA Kit for detecting AST in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P33097. Gene ID: 281206. Target Synonyms: cCAT; AAT; ASAT; SGOT; GOT1; Cysteine transaminase, cytoplasmic; Transaminase A; Aspartate Transaminase 1,Cytoplasmic; Glutamic-Oxaloacetic Transaminase 1,Soluble. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5241