Learn More
American Research Products Inc BoVine Beta HydroXybutyric Aci
Catalog No. 5000330112
Encompass
BoVine Beta-HydroXybutyric Acid b-OHB ELISA Kit from CUSABIO Detection Range 50 nmol/mL-800 nmol/mL Sensitivity 9 5 nmol/mL Sample serum plasma Method Competitive
Catalog No. 50-003-30112 Supplier American Research Products Inc Supplier No. CSBE10056B
BoVine Beta-HydroXybutyric Acid b-OHB ELISA Kit from CUSABIO Detection Range 50 nmol/mL-800 nmol/mL Sensitivity 9 5 nmol/mL Sample serum plasma Method Competitive