Learn More
American Research Products Inc BoVine beta lactoglobulin Bet
Catalog No. 5000327936
Encompass
BoVine beta-lactoglobulin (Beta-LG) (allergen Bos d 5) (LGB) ELISA kit from CUSABIO Detection Range 0 156 g/mL-10 g/mL Sensitivity 0 129 g/mL Sample serum plasma milk Method Competitive
Catalog No. 50-003-27936 Supplier American Research Products Inc Supplier No. CSBEL027168BO
BoVine beta-lactoglobulin (Beta-LG) (allergen Bos d 5) (LGB) ELISA kit from CUSABIO Detection Range 0 156 g/mL-10 g/mL Sensitivity 0 129 g/mL Sample serum plasma milk Method Competitive