Learn More
Hemostat Laboratories BOVINE BLOOD DEFIB 100ML
SDP
Catalog No. 50863769
Encompass_Preferred
Defibrinated Blood - Bovine 100Ml. Aseptically Collected From Healthy Animals Of U.S. Origin. Mechanically Defibrinated Without Anticoagulants Or Other Additives. Proprietary Screening Process Insures Low Fragility And Consistent Color.
Catalog No. 50-863-769 Supplier Hemostat Laboratories Supplier No. DBB100
Defibrinated Blood - Bovine 100Ml. Aseptically Collected From Healthy Animals Of U.S. Origin. Mechanically Defibrinated Without Anticoagulants Or Other Additives. Proprietary Screening Process Insures Low Fragility And Consistent Color.