Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Bovine Brain Extract

Catalog No. MP092850550 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
2.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP092850550 2.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP092850550 Supplier MP Biomedicals™ Supplier No. 092850550
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Filtered Bovine Brain Extract used for various endothelial cell type culture.

  • Bovine Brain Extract (BBE) is made from fresh, unfrozen pituitary glands from young, healthy cattle less than 30 months old. They are born, raised and slaughtered in New Zealand from a USDA approved source. New Zealand cattle are internationally recognized for being free from bovine spongiform encephalopathy (BSE, mad-cow disease) and have no history of blue tongue disease.
  • This Bovine Brain Extract (BBE) is formulated to be used at 0.1-0.2 % in the final serum-free medium. Studies have shown this level of BBE at the standard 7.5-15 micrograms/ mL will fully replace endothelial cell growth supplement (ECGS). BBE allows culture of these cells without feeder layers.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C, do not thaw and refreeze
Form Liquid
Sterility Sterile
For Use With (Application) Cell Culture
Quantity 2.5 mL
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.