Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Bovine Calf Serum, 1L, Iron-Fortified, United States Origin

Catalog No. 35055CM Shop All Corning Products
Change view
Click to view available options
Serum Treatment:
Gamma-irradiated
None
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Serum Treatment
35-055-CM Gamma-irradiated
35-053-CM None
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 35-055-CM Supplier Corning™ Supplier No. 35055CM
Only null left
Add to Cart
Add to Cart

Bovine calf serum is a cost-effective alternative to Fetal Bovine Serum (FBS). It is often used with robust cell lines when large quantities of serum are needed.

  • It is often used with robust cell lines when large quantities of serum are needed such as in the manufacturing of biological drugs and vaccines.
  • Bovine calf serum is derived from calves that are aged from 20 days up to 12 months.
  • It has very high levels of transferrin, which allows for iron fortification to levels of more than three times greater than normal FBS.
  • Iron is essential for cell respiration and metabolism and for some cell lines, makes up for the lower levels of growth factors and other metabolites found in calf serum when compared to FBS.
  • Sera Processing: Using industry leading techniques, blood is aseptically collected at abattoirs, centrifuged to isolate the serum and immediately frozen for testing and further processing. The raw sera is then true pooled and is triple 0.1 micron sterile filtered with a Sterility Assurance Level (SAL) of 10-3. Every lot then must pass USP <71> and EP 2.6.1 sterility testing, growth promotion testing, as well as test negative for mycoplasma and number of industry-monitored viruses. Additional testing includes endotoxin, hemoglobin, pH, osmolality, total protein, and a biochemical profile that contains various metals, proteins, hormones, and other metabolites.
  • EDQM Certificates of Suitability: This product has been granted a Certificate of Suitability, R0-CEP 2018-305, by the European Directorate for the Quality of Medicines (EDQM) certifying that the product meets the criteria described in the current version of the monograph no. 1483 of the European Pharmacopoeia “Product with risk of transmitting agent of animal spongiform encephalopathies
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -40°C to -10°C
Species Bovine, Bos taurus
Country of Origin United States
Form Liquid
Format Bottle
Product Type Calf Bovine Serum
Quantity Each
Serum Treatment Gamma-irradiated
Sterility Sterile
Sterilization Method Gamma-irradiated
Volume (Metric) 1 L
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.