Learn More
Sigma Aldrich Fine Chemicals Biosciences Bovine Calf Serum USA orig
Catalog No. 502906137
Encompass
Animal serum is commonly used to supplement basal media formulations for the optimal growth of many cell types in vitro. Bovine Calf Serum (BCS) is collected from calves that are less than 12 months old. The serum contains approximately double the protein of fetal bovine serum (fBS). BCS will promote growth in a variety of mammalian cell lines and is often used as a cost-effective substitute for fBS.
Catalog No. 50-290-6137 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 12133C1000ML
Animal serum is commonly used to supplement basal media formulations for the optimal growth of many cell types in vitro. Bovine Calf Serum (BCS) is collected from calves that are less than 12 months old. The serum contains approximately double the protein of fetal bovine serum (fBS). BCS will promote growth in a variety of mammalian cell lines and is often used as a cost-effective substitute for fBS.