Learn More
Biomatik Corporation Bovine Cathepsin D (CTSD) ELISA Kit
Supplier:  Biomatik Corporation EKU10710
Encompass_Preferred
Bovine Cathepsin D (CTSD) ELISA Kit for detecting CTSD in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.129ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P80209. Target Synonyms: CPSD; CLN10; Lysosomal Aspartyl Protease; Ceroid-Lipofuscinosis,Neuronal 10. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7790