Learn More
Biomatik Corporation Bovine Cholecystokinin (CCK) ELISA Kit
Supplier:  Biomatik Corporation EKU08697
Encompass_Preferred
Bovine Cholecystokinin (CCK) ELISA Kit for detecting CCK in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 12.35-1,000pg/ml. Sensitivity: 4.64pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P41520. Gene ID: 617510. Target Synonyms: CCK-PZ. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5983