Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Bovine chymotrypsinogen A native Protein from bovine pancreas, 6X crystallized

Catalog No. 89939615 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-939-615 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-939-615 Supplier Abnova™ Supplier No. P5334

Used for Enzyme, SDS-PAGE

Specifications

For Use With (Application) Enzyme Assay, SDS-PAGE
Name Bovine chymotrypsinogen A native Protein from bovine pancreas, 6X crystallized
Quantity 1 g
Storage Requirements Store at 4°C. This product is stable for at least 3 months.
Regulatory Status RUO
Common Name CHYMOTRYPSINOGEN
Biological Activity 72 units/mg material. 86 units/ mg protein. Intrinsic chymotrypsin activity: 0.275 units/mg. Unit Definition: When activated, one unit hydrolyzes one micromole of BTEE per minute at 25°C, pH 7.8.
Species Bovine
Protein Tag Untagged
Expression System Native protein purified from bovine pancreas
Form Lyophilized
Purity or Quality Grade ≥84%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.