Learn More
BioVendor, LLC Sircol; Bovine Collagen Reference Standard, 0.5 mg/ml, (3 x 5 ml), Biocolor Item # S1010
SDP
Supplier:  BioVendor, LLC S1010
Encompass
Sircol; Bovine Collagen Reference Standard, 0.5 mg/ml, (3 x 5 ml). Contains a sterile solution of cold acid-soluble collagen Type I, in 0.5M acetic acid within a sealed vial. Sircol™ Collagen Assay component. Biocolor Item # S1010
Catalog No. 50-172-5957