Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bovine Collagen Solution

Catalog No. 80461420ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
80-461-420ML 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 80-461-420ML Supplier Sigma Aldrich Supplier No. 80461420ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Bovine corium, acid solubilized collagen, telocollagen, pH 2 in 0.01 N HCl.
  • Predominantly Type I collagen with approximately 5% Type III, at 6 mg/ml.
  • Acid extracted collagen with telopeptide-intact.
  • Sterile filtered.
  • BSE-Free.
  • Forms a gel and is ideal for 3D biomaterial scaffolds for cell culture, engineering tissue, and drug delivery. Product of Collagen Solutions.

Specifications

Quantity 20 mL
Content And Storage 2°C to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.