Learn More
STEMCELL Technologies Collagen Solution, Size: 35 mL
SDP
Catalog No. NC9382691
Encompass
This product is suitable for the preparation of collagen gels for the culture of human hematopoietic progenitor cells using MegaCult™-C. This product can also be used to prepare collagen films (collagen coating) on cell culture surfaces.
Catalog No. NC9382691 Supplier STEMCELL Technologies Supplier No. 04902
This product is suitable for the preparation of collagen gels for the culture of human hematopoietic progenitor cells using MegaCult™-C. This product can also be used to prepare collagen films (collagen coating) on cell culture surfaces.