Learn More
American Research Products Inc BoVine complement fragment 5a
Catalog No. 5000327965
Encompass
BoVine complement fragment 5a (C5a) ELISA kit from CUSABIO Detection Range 4 69 ng/mL-300 ng/mL Sensitivity 1 17 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27965 Supplier American Research Products Inc Supplier No. CSBEQ027637BO
BoVine complement fragment 5a (C5a) ELISA kit from CUSABIO Detection Range 4 69 ng/mL-300 ng/mL Sensitivity 1 17 ng/mL Sample serum plasma tissue homogenates Method Sandwich