Learn More
American Research Products Inc BoVine Cu Zn SuperoXide Dismut
Catalog No. 5000326676
Encompass
BoVine Cu/Zn-SuperoXide Dismutase Cu/Zn-SOD ELISA Kit from CUSABIO Detection Range 15 6 ng/mL-1000 ng/mL Sensitivity 3 9 ng/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-26676 Supplier American Research Products Inc Supplier No. CSBE14090B
BoVine Cu/Zn-SuperoXide Dismutase Cu/Zn-SOD ELISA Kit from CUSABIO Detection Range 15 6 ng/mL-1000 ng/mL Sensitivity 3 9 ng/mL Sample serum plasma Method Sandwich